| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007357.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007357.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007357.hg.1 |
TC09000640.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008735.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008735.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008735.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008735.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008735.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008735.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007360.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007360.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007360.hg.1 |
TC09000640.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007356.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007356.hg.1 |
TC09000640.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900158238.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900158238.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900158238.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900158238.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900158238.hg.1 |
TC0900008735.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900008735.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008735.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008735.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008735.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008735.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008735.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900158237.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900158237.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900158237.hg.1 |
TC0900008735.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900158242.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900158242.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900158242.hg.1 |
ENST00000429139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900158242.hg.1 |
NR_038975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900158242.hg.1 |
TC0900008735.hg.1 |
Clariom D TC |
| Human Genome U95 Set |
58285_PM_at |
- |
- |
| Human Genome U95 Set |
58285_at |
- |
- |
| Human Genome U133 Set |
232478_PM_at |
- |
- |
| Human Genome U133 Set |
232478_at |
- |
- |