| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02025516.hg.1 |
NR_039627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02025516.hg.1 |
TC02001580.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011755.hg.1 |
ENST00000580105.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011755.hg.1 |
NR_039627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200185475.hg.1 |
ENST00000580105.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200185475.hg.1 |
NR_039627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200185475.hg.1 |
ENST00000580105.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200185475.hg.1 |
NR_039627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200185475.hg.1 |
TC0200011755.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200011755.hg.1 |
ENST00000580105.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011755.hg.1 |
NR_039627.1 |
Refseq |