Assay Details
| Gene Symbol: MIR4505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039727.1 | 1 | 41 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ELM2 and Myb/SANT domain containing 1 | ELMSAN1 |
C14orf117, C14orf43, LSR68, MIDEAS, c14_5541 |
Anti-sense | Chr. 14 - 73715122 - 73790285 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100506476 | LOC100506476 | 28.5kb | Sense | Chr. 14 - 73787360 - 73803628 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1400007656.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000449.hg.1
Set Membership: |
|

MIR4505
Genomic Map