| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000566575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
ENST00000582688.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008846.hg.1 |
NR_039760.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164084.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164084.hg.1 |
ENST00000582688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164084.hg.1 |
NR_039760.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164084.hg.1 |
TC2200008846.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164084.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164084.hg.1 |
ENST00000582688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164084.hg.1 |
NR_039760.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000291232.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000566575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
ENST00000582688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
NM_052945.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008846.hg.1 |
NR_039760.1 |
Refseq |