Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FOXP4 antisense RNA 1 | FOXP4-AS1 | -49.7kb | Anti-sense | Chr. 6 - 41523895 - 41548621 on NCBI Build GRCh38 | |
| Downstream | MyoD family inhibitor | MDFI |
I-MF, I-mfa |
38.2kb | Sense | Chr. 6 - 41637020 - 41654246 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map