| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008030.hg.1 |
ENST00000578459.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008030.hg.1 |
NR_039839.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11007968.hg.1 |
NR_039839.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11007968.hg.1 |
TC11000636.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1100008030.hg.1 |
ENST00000578459.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008030.hg.1 |
NR_039839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100158333.hg.1 |
ENST00000578459.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100158333.hg.1 |
NR_039839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100158333.hg.1 |
ENST00000578459.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100158333.hg.1 |
NR_039839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100158333.hg.1 |
TC1100008030.hg.1 |
Clariom D TC |