| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007514.hg.1 |
NR_039919.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007514.hg.1 |
TC22000386.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007629.hg.1 |
ENST00000585261.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007629.hg.1 |
NR_039919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200154253.hg.1 |
ENST00000585261.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200154253.hg.1 |
NR_039919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200154253.hg.1 |
TC2200007629.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200154253.hg.1 |
ENST00000585261.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200154253.hg.1 |
NR_039919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007629.hg.1 |
ENST00000585261.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007629.hg.1 |
NR_039919.1 |
Refseq |