Assay Details
| Gene Symbol: MIR4781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039942.1 | 1 | 40 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transcription elongation factor A N-terminal and central domain containing 2 | TCEANC2 | C1orf83 | Sense | Chr. 1 - 54053572 - 54112519 on NCBI Build GRCh38 | |
| Downstream | CUB domain containing protein 2 | CDCP2 | 84.8kb | Anti-sense | Chr. 1 - 54138995 - 54153006 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100008375.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000661.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01010726.hg.1 | NR_039942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010726.hg.1 | TC01000661.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000391366.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | ENST00000585250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NR_039942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000234827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000371331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000391366.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000498272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | ENST00000585250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NM_153035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NR_039942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008375.hg.1 | NR_130900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159638.hg.1 | ENST00000585250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159638.hg.1 | NR_039942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159638.hg.1 | ENST00000585250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159638.hg.1 | NR_039942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159638.hg.1 | TC0100008375.hg.1 | Clariom D TC |

MIR4781
Genomic Map