| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020821.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020821.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020821.hg.1 |
TC0X001552.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
ENST00000444722.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
ENST00000444722.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
ENST00000444722.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011241.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000452506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000452506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020820.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020820.hg.1 |
ENST00000444722.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020820.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020820.hg.1 |
TC0X001552.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00160927.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160927.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00160927.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160927.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00160927.hg.1 |
TC0X00008878.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000452506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000452506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
ENST00000444722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
ENST00000444722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
ENST00000444722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011241.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00160928.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160928.hg.1 |
ENST00000452506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160928.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00160928.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160928.hg.1 |
ENST00000452506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160928.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00160928.hg.1 |
TC0X00008878.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00176901.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00176901.hg.1 |
ENST00000444722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00176901.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176901.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00176901.hg.1 |
ENST00000444722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00176901.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176901.hg.1 |
TC0X00011241.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00176902.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00176902.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176902.hg.1 |
ENST00000412436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00176902.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176902.hg.1 |
TC0X00011241.hg.1 |
Clariom D TC |