| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006781.hg.1 |
ENST00000516515.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006901.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006901.hg.1 |
TC19000519.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009081.hg.1 |
ENST00000516302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012175.hg.1 |
ENST00000516372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010453.hg.1 |
ENST00000630438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006902.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006902.hg.1 |
TC19000519.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006903.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006903.hg.1 |
TC19000519.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006781.hg.1 |
ENST00000516515.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012175.hg.1 |
ENST00000516372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
TC1900007984.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159781.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009081.hg.1 |
ENST00000516302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010453.hg.1 |
ENST00000630438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
TC1900007984.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159782.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000586819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000587248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007984.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
TC1900007984.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159775.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
NR_040013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
TC1900007984.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000589653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000590433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
ENST00000592103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159776.hg.1 |
NR_040013.1 |
Refseq |