Assay Details
| Gene Symbol: LOC100631378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040015.1 | 3 - 4 | 271 | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC644554 | LOC644554 | -6.7kb | Anti-sense | Chr. 19 - 37817411 - 37826638 on NCBI Build GRCh38 | |
| Downstream | WD repeat domain 87 | WDR87 | NYD-SP11 | 47.6kb | Sense | Chr. 19 - 37884577 - 37906706 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001490.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19020236.hg.1 | ENST00000433142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19020236.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19020236.hg.1 | TC19001490.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | ENST00000592714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | ENST00000592714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010581.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | TC1900010581.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000587395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187018.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | NR_040015.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | TC1900010581.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | ENST00000433142.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | ENST00000592640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900187017.hg.1 | NR_040015.1 | Refseq |

LOC100631378
Genomic Map