| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
ENST00000592714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020235.hg.1 |
ENST00000433142.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020235.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020235.hg.1 |
TC19001490.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000433142.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000587395.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
ENST00000592714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010581.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900187015.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900187015.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900187015.hg.1 |
NR_040015.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900187015.hg.1 |
TC1900010581.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900187015.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900187015.hg.1 |
ENST00000592640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900187015.hg.1 |
NR_040015.1 |
Refseq |