| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19009350.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19009350.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19009350.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19009350.hg.1 |
TC19000666.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19009349.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19009349.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19009349.hg.1 |
TC19000666.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525352.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525352.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164423.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164423.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164423.hg.1 |
TC1900008386.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900164423.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164423.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164424.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164424.hg.1 |
ENST00000525352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164424.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164424.hg.1 |
TC1900008386.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900164424.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164424.hg.1 |
ENST00000525352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164424.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164422.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164422.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164422.hg.1 |
TC1900008386.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900164422.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164422.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164425.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164425.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164425.hg.1 |
TC1900008386.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900164425.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164425.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
TC1900008386.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164426.hg.1 |
NR_040042.1 |
Refseq |