| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525352.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
ENST00000525352.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19009351.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19009351.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19009351.hg.1 |
TC19000666.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900164429.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164429.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164429.hg.1 |
TC1900008386.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900164429.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164429.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
ENST00000525352.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008386.hg.1 |
NR_040042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164428.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164428.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164428.hg.1 |
TC1900008386.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900164428.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164428.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164427.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164427.hg.1 |
NR_040041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900164427.hg.1 |
TC1900008386.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900164427.hg.1 |
ENST00000500689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900164427.hg.1 |
NR_040041.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
1561532_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1561532_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561532_at2 |
- |
- |