Assay Details
| Gene Symbol: SDCBP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040048.1 | 1 - 2 | 185 | 75 | ||
| NR_040049.1 | 1 - 2 | 185 | 75 | |||
| GenBank mRNA | AK055993.1 | - | 1 - 2 | 107 | 75 | |
| BC034773.1 | - | 1 - 2 | 118 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FKBP1A-SDCBP2 readthrough (NMD candidate) | FKBP1A-SDCBP2 | Anti-sense | Chr. 20 - 1309909 - 1393172 on NCBI Build GRCh38 | ||
| Downstream | microRNA 6869 | MIR6869 | hsa-mir-6869 | 19.7kb | Anti-sense | Chr. 20 - 1392900 - 1392961 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000016.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20000201.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000201.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000201.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000201.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000201.hg.1 | TC20000016.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000412497.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000412497.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000607947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000607947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000607947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | ENST00000609745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000198.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000198.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000198.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000198.hg.1 | TC20000016.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000199.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000199.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000199.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000199.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000199.hg.1 | TC20000016.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000202.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000202.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000202.hg.1 | TC20000016.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000412497.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000412497.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000607947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000607947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000607947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | ENST00000609745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006486.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144429.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144429.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144429.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144429.hg.1 | TC2000006486.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000144429.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144429.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144429.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | TC2000006486.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144443.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000607947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000609745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | TC2000006486.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000607947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000609285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000609470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | ENST00000609745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | NR_040047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144433.hg.1 | NR_040049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144444.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144444.hg.1 | NR_040048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144444.hg.1 | TC2000006486.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000144444.hg.1 | ENST00000446423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144444.hg.1 | NR_040048.1 | Refseq |

SDCBP2-AS1
Genomic Map