| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015326.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015326.hg.1 |
ENST00000448377.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015326.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015326.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015326.hg.1 |
TC09001197.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000420573.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000420573.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000451488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000451488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000451488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015324.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015324.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015324.hg.1 |
TC09001197.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015333.hg.1 |
ENST00000448377.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015333.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015333.hg.1 |
TC09001197.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015332.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015332.hg.1 |
ENST00000448377.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015332.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015332.hg.1 |
TC09001197.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900186189.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186189.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186189.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186189.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900186189.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186189.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186189.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186189.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900186189.hg.1 |
TC0900012242.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900186194.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186194.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186194.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900186194.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186194.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186194.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900186194.hg.1 |
TC0900012242.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000420573.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000420573.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000451488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000451488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000451488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
ENST00000535188.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012242.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900186195.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186195.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900186195.hg.1 |
ENST00000448377.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186195.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900186195.hg.1 |
TC0900012242.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900186186.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186186.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900186186.hg.1 |
ENST00000377178.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900186186.hg.1 |
NR_040076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900186186.hg.1 |
TC0900012242.hg.1 |
Clariom D TC |