Assay Details
| Gene Symbol: MAMDC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040076.1 | 3 - 4 | 458 | 88 | ||
| GenBank mRNA | BC043510.1 | - | 3 - 4 | 402 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 9 open reading frame 135 | C9orf135 | -246.9kb | Anti-sense | Chr. 9 - 69820804 - 69906237 on NCBI Build GRCh38 | |
| Downstream | SMC5 antisense RNA 1 (head to head) | SMC5-AS1 | MAMDC2-AS2 | 58.3kb | Sense | Chr. 9 - 70216059 - 70258874 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001197.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3233956 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4063 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3233956 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09015322.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015322.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015322.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015322.hg.1 | TC09001197.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000420573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000420573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015320.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015320.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015320.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015320.hg.1 | TC09001197.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015323.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015323.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015323.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015323.hg.1 | TC09001197.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015321.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015321.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015321.hg.1 | TC09001197.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900186182.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186182.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186182.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186182.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186182.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186182.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186182.hg.1 | TC0900012242.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900186183.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186183.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186183.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186183.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186183.hg.1 | TC0900012242.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900186184.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186184.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186184.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186184.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186184.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186184.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186184.hg.1 | TC0900012242.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000420573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000420573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186185.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186185.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186185.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186185.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186185.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186185.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186185.hg.1 | TC0900012242.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1559655_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559656_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559655_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559656_PM_a_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559656_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559655_at1 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037554 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037554 | - | - |

MAMDC2-AS1
Genomic Map