Assay Details
| Gene Symbol: LOC100133286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040084.1 | 3 - 4 | 384 | 115 | ||
| GenBank mRNA | AK303953.1 | - | 3 - 4 | 384 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SET domain containing 4 | SETD4 |
C21orf18, C21orf27 |
-8.8kb | Sense | Chr. 21 - 36034541 - 36060818 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105369306 | LOC105369306 | 29.5kb | Anti-sense | Chr. 21 - 36104434 - 36109692 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000429.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 34527 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 150909 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 100868 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 107498 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 42916 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 630462 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 291424 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 713662 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 72893 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 282397 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 908265 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 761704 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 716481 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 479640 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 726607 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 62260 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 68749 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 886493 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 696932 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 328555 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 75561 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 302800 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 360719 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 289501 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 222584 | AL713792.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 222584 | AM774227.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 222584 | NM_001102608.1 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 390812 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 281345 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 291171 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 157025 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 205236 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 19940 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 4866 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60054 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P21246492 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P21246474 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P21246467 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P21246483 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3235117 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03583009 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21246467 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03583014 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P41443754 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21246474 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21246480 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21246492 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03583015 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005069.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005069.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005069.hg.1 | TC21000429.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005070.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005070.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005070.hg.1 | TC21000429.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | ENST00000415147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | ENST00000415147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008063.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | ENST00000415147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | ENST00000415147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008063.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100154463.hg.1 | ENST00000415147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154463.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154463.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154463.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100154463.hg.1 | TC2100008063.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100154463.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100154463.hg.1 | ENST00000415147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154463.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154463.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154462.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154462.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154462.hg.1 | NR_040084.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100154462.hg.1 | TC2100008063.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100154462.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154462.hg.1 | ENST00000535199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154462.hg.1 | NR_040084.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A018844 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A018844 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3211055 | NR_040084.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3211055 | AK303953.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3274814 | NR_040084.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3274814 | AK303953.1 | human_transcriptome |

LOC100133286
Genomic Map