| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013572.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013572.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013572.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013572.hg.1 |
TC20000997.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013570.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013570.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013570.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013570.hg.1 |
TC20000997.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009664.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009664.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009664.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009664.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009664.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009664.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009664.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009664.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009664.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000167412.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167412.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167412.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000167412.hg.1 |
TC2000009664.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000167412.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167412.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167412.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009664.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009664.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009664.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009664.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009664.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009664.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009664.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009664.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009664.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000167411.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167411.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167411.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000167411.hg.1 |
TC2000009664.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000167411.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167411.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167411.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000167409.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167409.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167409.hg.1 |
NR_040513.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000167409.hg.1 |
TC2000009664.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000167409.hg.1 |
ENST00000443055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167409.hg.1 |
ENST00000451510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000167409.hg.1 |
NR_040513.1 |
Refseq |