Assay Details
| Gene Symbol: DSCAM-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038896.1 | 3 | 1402 | 86 | ||
| NR_038898.1 | 2 | 1044 | 86 | |||
| NR_038899.1 | 3 | 946 | 86 | |||
| NR_038900.1 | 2 | 915 | 86 | |||
| GenBank mRNA | AF401029.1 | - | 2 | 1044 | 86 | |
| AF401030.1 | - | 2 | 915 | 86 | ||
| AF401033.1 | - | 3 | 1396 | 86 | ||
| AF401035.1 | - | 3 | 946 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P198445 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 38843 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P198445 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P198445 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P198445 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21001765.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001765.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001765.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001765.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001765.hg.1 | TC21000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC21000181.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001764.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001764.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001764.hg.1 | TC21000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001767.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001767.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001767.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001767.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001767.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001767.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001767.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001767.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001767.hg.1 | TC21000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001766.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001766.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001766.hg.1 | TC21000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001762.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001762.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001762.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001762.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001762.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001762.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001762.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001762.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001762.hg.1 | TC21000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001763.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001763.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001763.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001763.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001763.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001763.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001763.hg.1 | TC21000181.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | TC2100007184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148436.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | TC2100007184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148441.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | TC2100007184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148438.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007184.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | TC2100007184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000422749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000444046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | NR_038899.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148435.hg.1 | NR_038900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148440.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148440.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148440.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148440.hg.1 | TC2100007184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100148440.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148440.hg.1 | ENST00000427451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148440.hg.1 | NR_038896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148437.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148437.hg.1 | NR_038898.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148437.hg.1 | TC2100007184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100148437.hg.1 | ENST00000455354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148437.hg.1 | NR_038898.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1562821_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562821_PM_a_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562821_a_at6 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019275 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019276 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019276 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019275 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 216399 | - | - |

DSCAM-AS1
Genomic Map