Assay Details
| Gene Symbol: FAM212B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038951.1 | 3 | 2073 | 106 | ||
| NR_038952.1 | 2 | 1918 | 106 | |||
| GenBank mRNA | AK092511.1 | - | 3 | 1959 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928718 | LOC101928718 | Anti-sense | Chr. 1 - 111745317 - 111755509 on NCBI Build GRCh38 | ||
| Downstream | DEAD-box helicase 20 | DDX20 |
DP103, GEMIN3 |
7.8kb | Sense | Chr. 1 - 111755568 - 111767577 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FAM212B-AS1
Genomic Map