Assay Details
| Gene Symbol: RCAN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251977.1 | NP_001238906.1 | 3 - 4 | 650 | 110 | |
| NM_001251978.1 | NP_001238907.1 | 3 - 4 | 564 | 110 | ||
| NM_001251979.1 | NP_001238908.1 | 3 - 4 | 542 | 110 | ||
| NM_001251983.1 | NP_001238912.1 | 2 - 3 | 429 | 110 | ||
| NM_001251984.1 | NP_001238913.1 | 2 - 3 | 288 | 110 | ||
| NM_013441.3 | NP_038469.1 | 3 - 4 | 683 | 110 | ||
| GenBank mRNA | AF176116.1 | - | 2 - 3 | 392 | 110 | |
| BC035854.1 | - | 3 - 4 | 683 | 5770066 | 110 | |
| BX105019.1 | - | 1 - 2 | 110 | 110 | ||
| HQ287726.1 | - | 2 - 3 | 212 | 110 | ||
| HQ317421.1 | - | 3 - 4 | 518 | 110 | ||
| HQ317422.1 | - | 2 - 3 | 264 | 110 | ||
| HQ317423.1 | - | 3 - 4 | 564 | 110 | ||
| HQ317426.1 | - | 2 - 3 | 433 | 110 | ||
| HQ317427.1 | - | 1 - 2 | 179 | 110 | ||
| JN228252.1 | - | 3 - 4 | 537 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the HQ287726.1, HQ317422.1, HQ317427.1, NM_001251983.1, NM_001251984.1 transcripts.
Set Membership: |
|

RCAN3
Genomic Map