Assay Details
| Gene Symbol: GMNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251989.1 | NP_001238918.1 | 4 - 5 | 488 | 120 | |
| NM_001251990.1 | NP_001238919.1 | 4 - 5 | 412 | 120 | ||
| NM_001251991.1 | NP_001238920.1 | 4 - 5 | 396 | 120 | ||
| NM_015895.4 | NP_056979.1 | 4 - 5 | 620 | 120 | ||
| XM_005249159.1 | XP_005249216.1 | 4 - 5 | 659 | 120 | ||
| XM_011514651.2 | XP_011512953.1 | 3 - 4 | 2714 | 120 | ||
| GenBank mRNA | AF067855.1 | - | 3 - 4 | 306 | 120 | |
| AK021685.1 | - | 4 - 5 | 488 | 120 | ||
| AK222992.1 | - | 3 - 4 | 279 | 120 | ||
| AL558330.3 | - | 4 - 5 | 603 | 120 | ||
| BC005185.1 | - | 4 - 5 | 543 | 4101771 | 120 | |
| BC005389.1 | - | 4 - 5 | 595 | 3996265 | 120 | |
| BG777134.1 | - | 4 - 5 | 369 | 121 | ||
| BM699599.1 | - | 4 - 5 | 394 | 120 | ||
| CB131959.1 | - | 1 - 2 | 110 | 120 | ||
| CB992796.1 | - | 4 - 5 | 507 | 120 | ||
More Information
Set Membership: |
|

GMNN
Genomic Map