Assay Details
| Gene Symbol: MIR489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030164.1 | 1 | 36 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 653 | MIR653 |
MIRN653, hsa-mir-653, mir-653 |
-1.1kb | Sense | Chr. 7 - 93482760 - 93482855 on NCBI Build GRCh38 |
| Downstream | microRNA 4652 | MIR4652 | 232.9kb | Anti-sense | Chr. 7 - 93716928 - 93717005 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0700011805.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001610.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869345 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869358 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869339 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869363 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869366 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869356 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869333 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869363 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869356 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869333 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869339 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869366 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869358 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869345 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25869330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05618301 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700011805.hg.1 | ENST00000384923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011805.hg.1 | NR_030164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024427.hg.1 | ENST00000384923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024427.hg.1 | NR_030164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024427.hg.1 | TC07001610.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700184355.hg.1 | ENST00000384923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184355.hg.1 | NR_030164.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700184355.hg.1 | ENST00000384923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184355.hg.1 | NR_030164.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700184355.hg.1 | TC0700011805.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700011805.hg.1 | ENST00000384923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011805.hg.1 | NR_030164.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308365 | NR_030164.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308365 | NR_030164.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308365 | NR_030164.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308365 | NR_030164.1 | human_transcriptome |

MIR489
Genomic Map