| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000541673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000541673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000541673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000541673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000541673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12014524.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12014524.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12014524.hg.1 |
TC12001098.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000541673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000541673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000541673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000541673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000541673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
ENST00000544517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009608.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200171837.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171837.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171837.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200171837.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171837.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171837.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200171837.hg.1 |
TC1200009608.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200171836.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171836.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171836.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200171836.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171836.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171836.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200171836.hg.1 |
TC1200009608.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200171838.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171838.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171838.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200171838.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171838.hg.1 |
ENST00000501371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200171838.hg.1 |
NR_045725.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200171838.hg.1 |
TC1200009608.hg.1 |
Clariom D TC |