Assay Details
| Gene Symbol: CELF2-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046108.1 | 6 - 7 | 2120 | 130 | ||
| GenBank mRNA | BC046381.1 | - | 6 - 7 | 2122 | 130 | |
| DB336852.1 | - | 1 - 2 | 130 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 710 | LINC00710 | -119.4kb | Sense | Chr. 10 - 10934940 - 10952163 on NCBI Build GRCh38 | |
| Downstream | CELF2 antisense RNA 1 | CELF2-AS1 | C10orf31 | 244.0kb | Sense | Chr. 10 - 11316833 - 11344674 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001034.hg.1
Set Membership: |
|

CELF2-AS2
Genomic Map