| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009795.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009795.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009795.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009795.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009795.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009795.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165252.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300165252.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165252.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300165252.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165252.hg.1 |
TC1300009795.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300009795.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009795.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009795.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009795.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009795.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009795.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165254.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300165254.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165254.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300165254.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165254.hg.1 |
TC1300009795.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300165253.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300165253.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165253.hg.1 |
ENST00000417970.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300165253.hg.1 |
NR_046583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165253.hg.1 |
TC1300009795.hg.1 |
Clariom D TC |