| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000502373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000502373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000502373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000502373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000514364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000514364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
ENST00000514364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007524.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151499.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151499.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151499.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151499.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151499.hg.1 |
TC0400007524.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
ENST00000514364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
ENST00000514364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151498.hg.1 |
TC0400007524.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000502373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000502373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000502373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000502373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000514364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000514364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
ENST00000514364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007524.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151497.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151497.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151497.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151497.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151497.hg.1 |
ENST00000510785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151497.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151497.hg.1 |
ENST00000511989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151497.hg.1 |
NR_046622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151497.hg.1 |
TC0400007524.hg.1 |
Clariom D TC |