| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009337.hg.1 |
ENST00000457171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009337.hg.1 |
ENST00000457171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009337.hg.1 |
ENST00000457171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009337.hg.1 |
NR_046685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009337.hg.1 |
NR_046685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009337.hg.1 |
NR_046685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009337.hg.1 |
ENST00000457171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009337.hg.1 |
ENST00000457171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009337.hg.1 |
ENST00000457171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009337.hg.1 |
NR_046685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009337.hg.1 |
NR_046685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009337.hg.1 |
NR_046685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300162681.hg.1 |
ENST00000457171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300162681.hg.1 |
NR_046685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300162681.hg.1 |
ENST00000457171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300162681.hg.1 |
NR_046685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300162681.hg.1 |
TC1300009337.hg.1 |
Clariom D TC |