| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007230.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007230.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007230.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007230.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007230.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007230.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007230.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007230.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007230.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007230.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200150803.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200150803.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200150803.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200150803.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200150803.hg.1 |
TC1200007230.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200150807.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200150807.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200150807.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200150807.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200150807.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200150807.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200150807.hg.1 |
TC1200007230.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200150808.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200150808.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200150808.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200150808.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200150808.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200150808.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200150808.hg.1 |
TC1200007230.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200007230.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007230.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007230.hg.1 |
ENST00000536397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007230.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007230.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007230.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007230.hg.1 |
ENST00000537346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007230.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007230.hg.1 |
NR_046909.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007230.hg.1 |
NR_046909.1 |
Refseq |