| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000557625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000557625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000557625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
ENST00000557625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
NR_051990.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
NR_051990.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
NR_051990.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009163.hg.1 |
NR_051990.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000556617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000557625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000557625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000557625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
ENST00000557625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
NR_051990.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
NR_051990.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
NR_051990.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009163.hg.1 |
NR_051990.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400167642.hg.1 |
ENST00000557625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400167642.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400167642.hg.1 |
ENST00000557625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400167642.hg.1 |
NR_047477.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400167642.hg.1 |
TC1400009163.hg.1 |
Clariom D TC |