| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010919.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010919.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010919.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010919.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010919.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010919.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010919.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010919.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010919.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179557.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179557.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179557.hg.1 |
TC0300010919.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300179556.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179556.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179556.hg.1 |
TC0300010919.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300179559.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179559.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179559.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179559.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179559.hg.1 |
TC0300010919.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300179558.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179558.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179558.hg.1 |
ENST00000442534.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179558.hg.1 |
NR_048543.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179558.hg.1 |
TC0300010919.hg.1 |
Clariom D TC |