| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009472.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163399.hg.1 |
TC1300009472.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300163400.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163400.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163400.hg.1 |
TC1300009472.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163401.hg.1 |
TC1300009472.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000419288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_046520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009472.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163403.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163403.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163403.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163403.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163403.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163403.hg.1 |
ENST00000451897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163403.hg.1 |
NR_049775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163403.hg.1 |
NR_049776.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163403.hg.1 |
TC1300009472.hg.1 |
Clariom D TC |