Assay Details
| Gene Symbol: MIR5003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049799.1 | 1 | 42 | 77 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | Z18973.1 | - | Chr. - - [-] on Build | 77 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | translation machinery associated 7 homolog (S. cerevisiae) pseudogene | LOC100130394 | -5.4kb | Anti-sense | Chr. 5 - 172656359 - 172656734 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928093 | LOC101928093 | 93.2kb | Sense | Chr. 5 - 172755503 - 172762562 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0500009460.hg.1
Set Membership: |
|

MIR5003
Genomic Map