Assay Details
| Gene Symbol: MIR5188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049820.1 | 1 | 59 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin C | UBC | HMG20 | -506.0 | Anti-sense | Chr. 12 - 124911645 - 124915041 on NCBI Build GRCh38 |
| Downstream | transfer RNA-Phe (GAA) 1-4 | TRF-GAA1-4 |
TRF-GAA1-2, TRNAF2 |
12.2kb | Anti-sense | Chr. 12 - 124927843 - 124927915 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1200009298.hg.1
Set Membership: |
|

MIR5188
Genomic Map