Assay Details
| Gene Symbol: MIR5191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049823.1 | 1 | 55 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | IPO9 antisense RNA 1 | IPO9-AS1 | Anti-sense | Chr. 1 - 201688256 - 201829559 on NCBI Build GRCh38 | ||
| Downstream | microRNA 1231 | MIR1231 |
MIRN1231, hsa-mir-1231 |
89.0kb | Sense | Chr. 1 - 201808611 - 201808702 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100011189.hg.1
Set Membership: |
|

MIR5191
Genomic Map