| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009408.hg.1 |
ENST00000431460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009408.hg.1 |
ENST00000431460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009408.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009408.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009408.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165797.hg.1 |
ENST00000431460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165797.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165797.hg.1 |
TC2000009408.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000165797.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165797.hg.1 |
ENST00000431460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009408.hg.1 |
ENST00000431460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009408.hg.1 |
ENST00000431460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009408.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009408.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009408.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165796.hg.1 |
ENST00000431460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165796.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165796.hg.1 |
TC2000009408.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000165796.hg.1 |
ENST00000431460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165796.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165798.hg.1 |
NR_051976.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165798.hg.1 |
TC2000009408.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000165798.hg.1 |
NR_051976.1 |
Refseq |