Assay Details
| Gene Symbol: EIF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_052024.1 | 2 - 3 | 133 | 63 | ||
| GenBank mRNA | BG283650.1 | - | 1 - 2 | 126 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MMP24 antisense RNA 1 | MMP24-AS1 | -1.8kb | Sense | Chr. 20 - 35276333 - 35278157 on NCBI Build GRCh38 | |
| Downstream | FAM83C antisense RNA 1 | FAM83C-AS1 |
C20orf120, NCRNA00154, dJ614O4.3 |
759.0 | Anti-sense | Chr. 20 - 35285251 - 35285772 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal subunit export from nucleus
maturation of 5.8S rRNA
maturation of LSU-rRNA
regulation of glycolytic process
translational initiation
response to insulin
gene silencing by miRNA
miRNA mediated inhibition of translation
mature ribosome assembly
regulation of fatty acid biosynthetic process
regulation of megakaryocyte differentiation
positive regulation of translation
assembly of large subunit precursor of preribosome
regulation of reactive oxygen species metabolic process
translation initiation factor activity
protein binding
ribosome binding
ribosomal large subunit binding
ribosomal subunit export from nucleus
maturation of 5.8S rRNA
maturation of LSU-rRNA
regulation of glycolytic process
translational initiation
response to insulin
gene silencing by miRNA
miRNA mediated inhibition of translation
mature ribosome assembly
regulation of fatty acid biosynthetic process
regulation of megakaryocyte differentiation
positive regulation of translation
assembly of large subunit precursor of preribosome
regulation of reactive oxygen species metabolic process
translation initiation factor activity
protein binding
ribosome binding
ribosomal large subunit binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000374436.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000374443.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000374450.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000440766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000447927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000621148.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NM_001267810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NM_002212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NM_181466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NM_181468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NR_052022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NR_052023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NR_052024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | TC2000010006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000374436.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000374443.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000374450.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000440766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000447927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | ENST00000621148.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NM_001267810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NM_002212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NM_181466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NM_181468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NR_052022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NR_052023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171468.hg.1 | NR_052024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000374436.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000374443.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000374450.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000440766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000447927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000621148.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NM_001267810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NM_002212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NM_181466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NM_181468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NR_052022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NR_052023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NR_052024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | TC2000010006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000374436.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000374443.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000374450.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000440766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000447927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | ENST00000621148.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NM_001267810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NM_002212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NM_181466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NM_181468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NR_052022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NR_052023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171465.hg.1 | NR_052024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000374436.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000374443.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000374450.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000440766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000447927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000621148.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NM_001267810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NM_002212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NM_181466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NM_181468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NR_052022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NR_052023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NR_052024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | TC2000010006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000374436.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000374443.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000374450.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000440766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000447927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | ENST00000621148.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NM_001267810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NM_002212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NM_181466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NM_181468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NR_052022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NR_052023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171466.hg.1 | NR_052024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000374436.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000374443.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000374450.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000415116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000440766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000447927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000621148.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NM_001267810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NM_002212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NM_181466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NM_181468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NR_052022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NR_052023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NR_052024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | TC2000010006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000374436.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000374443.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000374450.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000415116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000440766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000447927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | ENST00000621148.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NM_001267810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NM_002212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NM_181466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NM_181468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NR_052022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NR_052023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171470.hg.1 | NR_052024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000374436.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000374443.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000374450.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000415116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000440766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000447927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000462894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000621148.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NM_001267810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NM_002212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NM_181466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NM_181468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NR_052022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NR_052023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NR_052024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | TC2000010006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000374436.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000374443.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000374450.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000415116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000440766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000447927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000462894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | ENST00000621148.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NM_001267810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NM_002212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NM_181466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NM_181468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NR_052022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NR_052023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171471.hg.1 | NR_052024.1 | Refseq |

EIF6
Genomic Map