Assay Details
| Gene Symbol: ENKUR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_072993.1 | 2 - 3 | 445 | 124 | ||
| GenBank mRNA | DA285403.1 | - | 2 - 3 | 372 | 124 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphoribosyl transferase domain containing 1 | PRTFDC1 | HHGP | -28.2kb | Sense | Chr. 10 - 24848607 - 24952644 on NCBI Build GRCh38 |
| Downstream | threonine synthase like 1 | THNSL1 | TSH1 | 26.0kb | Anti-sense | Chr. 10 - 24952319 - 25026664 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168623.hg.1 | TC1000010071.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168624.hg.1 | TC1000010071.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |

ENKUR
Genomic Map