Assay Details
| Gene Symbol: NRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270707.1 | NP_001257636.1 | 3 - 4 | 673 | 76 | |
| NM_007243.2 | NP_009174.1 | 4 - 5 | 852 | 76 | ||
| NR_073065.1 | 3 - 4 | 677 | 76 | |||
| NR_073066.1 | 2 - 3 | 458 | 76 | |||
| XM_017010224.1 | XP_016865713.1 | 3 - 4 | 854 | 76 | ||
| XM_017010225.1 | XP_016865714.1 | 4 - 5 | 768 | 76 | ||
| GenBank mRNA | AF143676.1 | - | 3 - 4 | 498 | 76 | |
| AK075509.1 | - | 4 - 5 | 854 | 76 | ||
| AK298152.1 | - | 2 - 3 | 315 | 76 | ||
| AK315190.1 | - | 3 - 4 | 510 | 76 | ||
| AY358411.1 | - | 3 - 4 | 519 | 76 | ||
| BC004509.2 | - | 1 - 2 | 124 | 76 | ||
| BC021260.1 | - | 3 - 4 | 490 | 76 | ||
| BC039865.1 | - | 3 - 4 | 518 | 6043663 | 76 | |
| BE548635.1 | - | 3 - 4 | 498 | 76 | ||
| BU159435.1 | - | 3 - 4 | 482 | 76 | ||
More Information
Set Membership: |
|

NRM
Genomic Map