Assay Details
| Gene Symbol: BCL2L13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270734.1 | NP_001257663.1 | 4 - 5 | 738 | 70 | |
| NR_073068.1 | 4 - 5 | 550 | 70 | |||
| NR_073069.1 | 4 - 5 | 533 | 70 | |||
| XM_011546123.1 | XP_011544425.1 | 5 - 6 | 680 | 70 | ||
| GenBank mRNA | AF539453.1 | - | 4 - 5 | 483 | 70 | |
| AL831982.2 | - | 4 - 5 | 512 | 70 | ||
| CN420914.1 | - | 4 - 5 | 532 | 70 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_011546123.1 transcripts.
Set Membership: |
|

BCL2L13
Genomic Map