Assay Details
| Gene Symbol: GNPDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073094.1 | 4 - 5 | 513 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
GUF1 homolog, GTPase |
GUF1 | EF-4 | -10.0kb | Anti-sense | Chr. 4 - 44678395 - 44700685 on NCBI Build GRCh38 |
| Downstream | gamma-aminobutyric acid type A receptor gamma1 subunit | GABRG1 | 1.3mb | Sense | Chr. 4 - 46035769 - 46124065 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

GNPDA2
Genomic Map