Assay Details
| Gene Symbol: NHLRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012754.3 | NP_001012772.1 | 5 - 6 | 1009 | 85 | |
| NM_001017370.2 | NP_001017370.1 | 4 - 5 | 808 | 85 | ||
| NR_073109.1 | 5 - 6 | 751 | 85 | |||
| GenBank mRNA | AK299198.1 | - | 5 - 6 | 922 | 85 | |
| AK300260.1 | - | 4 - 5 | 721 | 85 | ||
| BC052492.1 | - | 5 - 6 | 689 | 85 | ||
| BC140877.1 | - | 5 - 6 | 935 | 9021745 | 85 | |
| BC140878.1 | - | 5 - 6 | 935 | 9021746 | 85 | |
| BX647872.1 | - | 4 - 5 | 698 | 85 | ||
| BX648122.1 | - | 5 - 6 | 961 | 85 | ||
| CR749318.1 | - | 4 - 5 | 730 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000135.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000485407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000485407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000485407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000485407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001566.hg.1 | ENST00000379599.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001566.hg.1 | ENST00000379600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001566.hg.1 | ENST00000470258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001566.hg.1 | ENST00000537150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001566.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001566.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001566.hg.1 | TC13000135.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147050.hg.1 | TC1300006923.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379599.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000379600.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000470258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000473371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000485407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000485407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000485407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000485407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | ENST00000610563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001012754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NM_001017370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NR_073109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006923.hg.1 | NR_073109.1 | Refseq |

NHLRC3
Genomic Map