Assay Details
| Gene Symbol: NHLRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012754.3 | NP_001012772.1 | 6 - 7 | 1118 | 122 | |
| NM_001017370.2 | NP_001017370.1 | 5 - 6 | 917 | 122 | ||
| NR_073109.1 | 6 - 7 | 860 | 122 | |||
| GenBank mRNA | AK299198.1 | - | 6 - 7 | 1031 | 122 | |
| AK300260.1 | - | 5 - 6 | 830 | 122 | ||
| BC052492.1 | - | 6 - 7 | 798 | 122 | ||
| BC140877.1 | - | 6 - 7 | 1044 | 9021745 | 122 | |
| BC140878.1 | - | 6 - 7 | 1044 | 9021746 | 122 | |
| BX647872.1 | - | 5 - 6 | 807 | 122 | ||
| BX648122.1 | - | 6 - 7 | 1070 | 122 | ||
| CR749318.1 | - | 5 - 6 | 839 | 122 | ||
More Information
Set Membership: |
|

NHLRC3
Genomic Map