Assay Details
| Gene Symbol: CHEK2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038836.1 | 4 - 5 | 563 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
BMS1, ribosome biogenesis factor pseudogene 15 |
BMS1P15 | -124.4kb | Sense | Chr. 15 - 20153776 - 20160826 on NCBI Build GRCh38 | |
| Downstream | hect domain and RLD 2 pseudogene 3 | HERC2P3 | D15F37S4 | 118.2kb | Anti-sense | Chr. 15 - 20408396 - 20506180 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000003.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P109452 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15000005.hg.1 | ENST00000555186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000005.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000005.hg.1 | TC15000003.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000006.hg.1 | ENST00000555186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000006.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000006.hg.1 | TC15000003.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000009.hg.1 | ENST00000555186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000009.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000009.hg.1 | TC15000003.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000010.hg.1 | ENST00000555186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000010.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000010.hg.1 | TC15000003.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000568520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000568520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000568520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000568520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010094.hg.1 | ENST00000568520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000008.hg.1 | ENST00000555186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000008.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000008.hg.1 | TC15000003.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000007.hg.1 | ENST00000555186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000007.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000007.hg.1 | TC15000003.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000567422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000568520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000568520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000568520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000568520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010094.hg.1 | ENST00000568520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144107.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144107.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144107.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144107.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144107.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144107.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144107.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144107.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144107.hg.1 | TC1500006447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500144119.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144119.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144119.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144119.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144119.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144119.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144119.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144119.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144119.hg.1 | TC1500006447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500144108.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144108.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144108.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144108.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144108.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144108.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144108.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144108.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144108.hg.1 | TC1500006447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500144100.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144100.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144100.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144100.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144100.hg.1 | TC1500006447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500144104.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144104.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144104.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144104.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144104.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144104.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144104.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144104.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144104.hg.1 | TC1500006447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500144117.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144117.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144117.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144117.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144117.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144117.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144117.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144117.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144117.hg.1 | TC1500006447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006447.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144103.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144103.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144103.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144103.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144103.hg.1 | ENST00000554701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144103.hg.1 | ENST00000555186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144103.hg.1 | ENST00000566448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500144103.hg.1 | NR_038836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500144103.hg.1 | TC1500006447.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052623 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052623 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1736346 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1736346 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1736346 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1736346 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1736346 | - | - |

CHEK2P2
Genomic Map