Assay Details
| Gene Symbol: MIR767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030409.1 | 1 | 44 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 105-1 | MIR105-1 |
MIRN105-1, mir-105-1 |
-1.1kb | Sense | Chr. X - 152392219 - 152392299 on NCBI Build GRCh38 |
| Downstream | microRNA 105-2 | MIR105-2 |
MIRN105-2, mir-105-2 |
883.0 | Sense | Chr. X - 152394412 - 152394492 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0X00011129.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001498.hg.1
Set Membership: |
|

MIR767
Genomic Map