Assay Details
| Gene Symbol: MIR1237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031602.1 | 1 | 50 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 88B | CCDC88B |
BRLZ, CCDC88, HKRP3, gipie |
-11.1kb | Sense | Chr. 11 - 64340203 - 64357534 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100996455 | LOC100996455 | 80.4kb | Sense | Chr. 11 - 64449057 - 64451657 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1100007953.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000600.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838661 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28251003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16460976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16460978 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28251032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28251025 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838683 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838654 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16460979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838649 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838648 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28251034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16460983 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838688 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838663 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16460978 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28251034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838663 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16460976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28251003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838688 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16460979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28251025 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28251032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838649 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838648 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838683 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16460983 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838654 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838661 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838671 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007953.hg.1 | ENST00000408346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007953.hg.1 | NR_031602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007437.hg.1 | ENST00000408346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007437.hg.1 | NR_031602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007437.hg.1 | TC11000600.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100007953.hg.1 | ENST00000408346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007953.hg.1 | NR_031602.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157160.hg.1 | ENST00000408346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157160.hg.1 | NR_031602.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157160.hg.1 | ENST00000408346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157160.hg.1 | NR_031602.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157160.hg.1 | TC1100007953.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309041 | NR_031602.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309041 | NR_031602.1 | human_transcriptome |

MIR1237
Genomic Map