| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007133.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007133.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007133.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007133.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007133.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007133.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300148297.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300148297.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300148297.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300148297.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300148297.hg.1 |
TC1300007133.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300007133.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007133.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007133.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007133.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007133.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007133.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300148298.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300148298.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300148298.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300148298.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300148298.hg.1 |
TC1300007133.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300148299.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300148299.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300148299.hg.1 |
ENST00000422483.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300148299.hg.1 |
NR_046511.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300148299.hg.1 |
TC1300007133.hg.1 |
Clariom D TC |