Assay Details
| Gene Symbol: RPL13AP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003932.2 | 1 | 231 | 112 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DEAD-box helicase 47 | DDX47 |
E4-DBP, HQ0256, MSTP162, RRP3 |
-45.5kb | Sense | Chr. 12 - 12813346 - 12829981 on NCBI Build GRCh38 |
| Downstream | G protein-coupled receptor class C group 5 member A | GPRC5A |
GPCR5A, PEIG-1, RAI3, RAIG1, TIG1 |
14.9kb | Sense | Chr. 12 - 12891022 - 12913666 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000181.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3340060 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P90143 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P272061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197542 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630929 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197546 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630900 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630914 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630908 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630918 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197538 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630905 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197543 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197551 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197540 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197529 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197548 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197551 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630914 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197542 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197538 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630918 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197548 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197529 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197546 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630929 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630900 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197540 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197543 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630908 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08197526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28630905 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197553 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197527 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197580 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197560 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197535 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16535457 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197548 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197518 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197585 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197542 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197544 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197538 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197528 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197521 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197586 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197584 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197546 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197578 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16535465 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197555 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16535468 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197531 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197557 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197550 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197540 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08197561 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8030357 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8030353 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8030355 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8030359 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7954064 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8030358 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7954063 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8030351 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002294.hg.1 | NR_003932.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002294.hg.1 | TC12000181.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200148195.hg.1 | NR_003932.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200148195.hg.1 | NR_003932.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200148195.hg.1 | TC1200006894.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200148194.hg.1 | ENST00000459725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200148194.hg.1 | NR_003932.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200148194.hg.1 | ENST00000459725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200148194.hg.1 | NR_003932.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200148194.hg.1 | TC1200006894.hg.1 | Clariom D TC |
| Human Genome U95 Set | 74560_f_at | - | - |
| Human Genome U95 Set | 74560_PM_f_at | - | - |
| Human Genome U95 Set | 67923_at | - | - |
| Human Genome U95 Set | 67923_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048392 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048392 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1674983 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1674983 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1674983 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1660376 | BC065236.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1660376 | BC001675.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1660376 | BC001836.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1660376 | BC013230.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1674983 | - | - |

RPL13AP20
Genomic Map