Assay Details
| Gene Symbol: TFPI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271003.1 | NP_001257932.1 | 3 - 4 | 746 | 91 | |
| NM_006528.3 | NP_006519.1 | 3 - 4 | 779 | 91 | ||
| GenBank mRNA | AK024817.1 | - | 3 - 4 | 541 | 91 | |
| AK075427.1 | - | 3 | 540 | 142 | ||
| AK092499.1 | - | 3 - 4 | 508 | 91 | ||
| AK129833.1 | - | 3 - 4 | 541 | 91 | ||
| AK313260.1 | - | 3 - 4 | 541 | 91 | ||
| AY691946.1 | - | 3 - 4 | 517 | 91 | ||
| BC005330.1 | - | 3 - 4 | 541 | 3929933 | 91 | |
| BC034155.1 | - | 2 - 3 | 264 | 91 | ||
| D29992.1 | - | 3 - 4 | 522 | 91 | ||
| L27624.1 | - | 3 - 4 | 504 | 91 | ||
More Information
Set Membership: |
|

TFPI2
Genomic Map